5-Bromo-2-benzoxazolinone 97%
Supplier: Ambeed
|
Warning
|
A157614-10G
A157614-250MG
A157614-25G
A157614-1G
A157614-5G
103805-024EA
12
USD
103805-024
103843-226
76768-428
76768-430
77300-274
77319-060
77319-062
5-Bromo-2-benzoxazolinone 97%
5-Bromo-2-benzoxazolinone
Precaución: for Research Use Only
Formula:
C₇H₄BrNO₂ Melting Pt: 218…222 °C Storage Temperature: Ambient |
MDL Number:
MFCD01664246 CAS Number: 14733-73-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...