5-Bromobenzo[d]isoxazole 98%

Supplier: Ambeed
Warning

A239442-100MG A239442-250MG A239442-5G
103811-290EA 63 USD
103811-290 76786-478 76786-480 76786-482
5-Bromobenzo[d]isoxazole 98%
5-Bromobenzo[d]isoxazole

Precaución: for Research Use Only

Formula: C₇H₄BrNO
MW: 198.0200 g/mol
Flash Pt: 120.794 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06659644
CAS Number: 837392-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR