5-Bromobenzo[d]isoxazol-3(2H)-one 95%

Supplier: Ambeed
Warning

A189281-100MG A189281-250MG A189281-1G
103816-176EA 123 USD
103816-176 76813-862 76813-864 77084-538
5-Bromobenzo[d]isoxazol-3(2H)-one 95%
5-Bromobenzo[d]isoxazol-3(2H)-one

Precaución: for Research Use Only

Formula: C₇H₄BrNO₂
MW: 214.02 g/mol
Boiling Pt: 360.6 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD01658969
CAS Number: 65685-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR