5-Bromobenzo[b]thiophene-2-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
A578814-10G
A578814-250MG
A578814-1G
A578814-25G
A578814-5G
103828-852EA
8
USD
103828-852
103840-000
76775-048
76775-050
76844-128
76844-130
76844-132
5-Bromobenzo[b]thiophene-2-carboxylic acid 95%
5-Bromobenzo[b]thiophene-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₉H₅BrO₂S Boiling Pt: 429.7 °C (760 mm Hg) Melting Pt: 233…234 °C Flash Pt: 213.7 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01929338 CAS Number: 7312-10-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...