5-Bromoresorcinol 97%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-3,5-dihydroxybenzene, 5-Bromobenzene-1,3-diol, 3,5-Dihydroxybromobenzene

A122893-100MG A122893-10G A122893-250MG A122893-25G A122893-1G A122893-5G A122893-100G
103800-300EA 11 USD
103800-300 77026-560 77026-562 77026-564 77026-566 77312-420 77312-422 77566-342
5-Bromoresorcinol 97%
5-Bromoresorcinol

Precaución: For research use only.

Formula: C₆H₅BrO₂
MW: 189.01 g/mol
Boiling Pt: 299.9 °C (760 mm Hg)
Flash Pt: 135.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16293393
CAS Number: 106120-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR