5-Bromobenzene-1,3-diamine 95%

Supplier: Ambeed
Warning

Synonyms: 5-Bromophenylene-1,3-diamine, 1-Bromo-3,5-diaminobenzene

A244586-100MG A244586-10G A244586-250MG A244586-5G
103811-590EA 38 USD
103811-590 103851-422 76987-714 76987-716 76987-718 77090-632
5-Bromobenzene-1,3-diamine 95%
5-Bromobenzene-1,3-diamine

Precaución: for Research Use Only

Formula: C₆H₇BrN₂
MW: 187.037 g/mol
Boiling Pt: 336.144 °C (760 mm Hg)
Flash Pt: 157.094 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09036306
CAS Number: 33786-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR