5-Bromo-8-nitroquinoline 95%

Supplier: Ambeed
Warning

A276732-100MG A276732-250MG A276732-1G
103813-320EA 60 USD
103813-320 103853-256 77032-058 77032-060 77305-250
5-Bromo-8-nitroquinoline 95%
5-Bromo-8-nitroquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrN₂O₂
MW: 253.05 g/mol
Boiling Pt: 370.5 °C (760 mm Hg)
Flash Pt: 177.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00466078
CAS Number: 176967-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR