5-Bromo-8-methylquinoline 97%

Supplier: Ambeed
Warning

A482085-250MG A482085-1G A482085-5G
103828-140EA 17 USD
103828-140 103839-216 76830-004 76843-158 76843-160
5-Bromo-8-methylquinoline 97%
5-Bromo-8-methylquinoline

Precaución: for Research Use Only

Formula: C₁₀H₈BrN
MW: 222.081 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04966995
CAS Number: 74316-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR