5-Bromo-8-methoxy-2-methylquinoline 98%

Supplier: Ambeed
Warning

A121725-250MG A121725-1G A121725-100MG
103800-118EA 274 USD
103800-118 76971-552 77312-248 77880-986
5-Bromo-8-methoxy-2-methylquinoline 98%
5-Bromo-8-methoxy-2-methylquinoline

Precaución: for Research Use Only

Formula: C₁₁H₁₀BrNO
Boiling Pt: 341 °C (760 mm Hg)
Melting Pt: 114 °C
Flash Pt: 160 °C
Storage Temperature: Ambient
MDL Number: MFCD04966993
CAS Number: 103862-55-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR