5-Bromo-8-chloroquinoline 98%

Supplier: Ambeed
Warning

A239985-250MG A239985-1G A239985-5G
103811-324EA 20 USD
103811-324 103851-122 77113-242 77304-178 77304-180
5-Bromo-8-chloroquinoline 98%
5-Bromo-8-chloroquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrClN
Storage Temperature: Ambient
MDL Number: MFCD07700234
CAS Number: 927800-41-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR