5-Bromo-7-nitroindoline 97%

Supplier: Ambeed
Danger

A394432-10G A394432-250MG A394432-1G A394432-25G A394432-5G
103826-944EA 43 USD
103826-944 103837-982 76773-662 76773-664 76841-394 76841-396 76841-398
5-Bromo-7-nitroindoline 97%
5-Bromo-7-nitroindoline

Precaución: for Research Use Only

Formula: C₈H₇BrN₂O₂
MW: 243.06 g/mol
Boiling Pt: 344.2 °C (760 mm Hg)
Melting Pt: 133…136 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005708
CAS Number: 80166-90-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR