5-Bromo-7-methyl-1H-indole 98%
Supplier: Ambeed
|
Warning
|
A113391-100MG
A113391-250MG
A113391-1G
A113391-5G
103798-224EA
41
USD
103798-224
103824-066
76971-006
76971-008
77297-240
77297-242
5-Bromo-7-methyl-1H-indole 98%
5-Bromo-7-methyl-1H-indole
Precaución: for Research Use Only
Formula:
C₉H₈BrN Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD12962648 CAS Number: 15936-81-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...