5-Bromo-7-fluoroindole 95%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-7-fluoro-1H-indole

A114688-100MG A114688-250MG
103798-448EA 78 USD
103798-448 103824-290 76995-444 76995-446
5-Bromo-7-fluoroindole 95%
5-Bromo-7-fluoroindole

Precaución: for Research Use Only

Formula: C₈H₅BrFN
MW: 214.04 g/mol
Boiling Pt: 315.1 °C (760 mm Hg)
Flash Pt: 144.3 °C
Storage Temperature: Ambient
MDL Number: MFCD04037876
CAS Number: 883500-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR