5-Bromo-6-trifluoromethylpyridin-2-ylamine 95%

Supplier: Ambeed
Warning

A454904-100MG A454904-10G A454904-250MG A454904-1G A454904-5G
103827-828EA 21 USD
103827-828 103838-898 76774-154 76774-156 76774-158 76842-744 76842-746
5-Bromo-6-trifluoromethylpyridin-2-ylamine 95%
5-Bromo-6-trifluoromethylpyridin-2-ylamine

Precaución: for Research Use Only

Formula: C₆H₄BrF₃N₂
MW: 241.0086 g/mol
Boiling Pt: 218.1 °C (760 mm Hg)
Flash Pt: 86 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11848010
CAS Number: 882500-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR