5-Bromo-6-hydroxynicotinonitrile 95%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-6-oxo-1,6-dihydropyridine-3-carbonitrile

A139895-100MG A139895-250MG
103802-790EA 56 USD
103802-790 76996-832 76996-834
5-Bromo-6-hydroxynicotinonitrile 95%
5-Bromo-6-hydroxynicotinonitrile

Precaución: for Research Use Only

Formula: C₆H₃BrN₂O
MW: 199.01 g/mol
Boiling Pt: 345.139 °C (760 mm Hg)
Melting Pt: 259…261 °C at N/A mmHg
Flash Pt: 162.534 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11100229
CAS Number: 19840-44-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR