5-Bromo-6-methylpyrimidine-2,4-diol 97%

Supplier: Ambeed
Warning

A105523-250MG A105523-25G A105523-5G A105523-1G
103796-896EA 34 USD
103796-896 76765-472 76765-474 76765-476 77309-500
5-Bromo-6-methylpyrimidine-2,4-diol 97%
5-Bromo-6-methylpyrimidine-2,4-diol

Precaución: for Research Use Only

Formula: C₅H₅BrN₂O₂
Melting Pt: 269…271 °C
Storage Temperature: Ambient
MDL Number: MFCD12964069
CAS Number: 15018-56-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR