5-Bromo-2-hydroxy-6-picoline 96%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-6-hydroxy-2-methylpyridine, 5-Bromo-6-methylpyridin-2-ol

A172610-1G A172610-25G A172610-5G A172610-100G A172610-10G
103806-732EA 6 USD
103806-732 103845-020 77082-222 77082-224 77082-226 77163-612 77163-614
5-Bromo-2-hydroxy-6-picoline 96%
5-Bromo-2-hydroxy-6-picoline

Precaución: for Research Use Only

Formula: C₆H₆BrNO
MW: 188.02 g/mol
Boiling Pt: 294.4 °C (760 mm Hg)
Melting Pt: 199…203 °C
Storage Temperature: Ambient
MDL Number: MFCD03427653
CAS Number: 54923-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR