5-Bromo-6-methylpyrazin-2-amine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-5-bromo-6-methylpyrazine

A353226-100MG A353226-250MG
103826-088EA 88 USD
103826-088 103837-090 76978-532 76978-534
5-Bromo-6-methylpyrazin-2-amine 98%
5-Bromo-6-methylpyrazin-2-amine

Precaución: for Research Use Only

Formula: C₅H₆BrN₃
MW: 188.0300 g/mol
Boiling Pt: 273.7 °C (760 mm Hg)
Flash Pt: 119.375 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08705763
CAS Number: 74290-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR