5-Bromo-6-methylpicolinonitrile 97%

Supplier: Ambeed
Warning

A177572-10G A177572-250MG A177572-1G
103807-242EA 15 USD
103807-242 103845-572 76824-780 76824-782 77082-960
5-Bromo-6-methylpicolinonitrile 97%
5-Bromo-6-methylpicolinonitrile

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
Boiling Pt: 273 °C (760 mm Hg)
Flash Pt: 119 °C
Storage Temperature: Ambient
MDL Number: MFCD13181587
CAS Number: 1173897-86-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR