5-Bromo-6-methylpyridine-2-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-6-methylpicolinic acid, 3-Bromo-2-methylpyridine-6-carboxylic acid

A123744-100G A123744-100MG A123744-10G A123744-250MG A123744-25G A123744-1G A123744-5G
103800-438EA 23 USD
103800-438 76810-684 76810-686 76810-688 76810-690 76810-692 77312-560 77312-562
5-Bromo-6-methylpyridine-2-carboxylic acid 95%
5-Bromo-6-methylpyridine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₆BrNO₂
MW: 216.032 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13181588
CAS Number: 137778-20-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR