5-Bromo-6-methyl-1H-indazole 97%
Supplier: Ambeed
|
Warning
|
A121792-100MG
A121792-10G
A121792-250MG
A121792-25G
A121792-1G
A121792-5G
103800-132EA
9
USD
103800-132
76766-528
76766-530
76766-532
76766-534
77312-260
77312-262
5-Bromo-6-methyl-1H-indazole 97%
5-Bromo-6-methyl-1H-indazole
Precaución: for Research Use Only
Formula:
C₈H₇BrN₂ MW: 211.0600 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09026991 CAS Number: 885223-72-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...