5-Bromo-6-methyl-1H-benzimidazole 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-6-methyl-1H-benzo[d]imidazole

A230284-250MG A230284-1G
103819-536EA 232 USD
103819-536 103850-434 76987-318 77089-292
5-Bromo-6-methyl-1H-benzimidazole 97%
5-Bromo-6-methyl-1H-benzimidazole

Precaución: for Research Use Only

Formula: C₈H₇BrN₂
MW: 211.059 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11846340
CAS Number: 116106-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR