5-Bromo-6-methoxypyridin-2-amine 98%
Supplier: Ambeed
|
Warning
|
A238856-100G
A238856-1G
A238856-250MG
103811-250EA
41
USD
103811-250
103851-046
76826-676
77090-182
77683-722
5-Bromo-6-methoxypyridin-2-amine 98%
5-Bromo-6-methoxypyridin-2-amine
Precaución: For research use only.
Formula:
C₆H₇BrN₂O MW: 203.04 g/mol Melting Pt: 79…84 °C Storage Temperature: Ambient |
MDL Number:
MFCD08062947 CAS Number: 1211533-83-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...