5-Bromo-6-methoxy-1H-pyrrolo[2,3-b]pyridine 97%

Supplier: Ambeed
Warning

A138123-100MG A138123-250MG A138123-5G A138123-1G
103802-556EA 83 USD
103802-556 76811-624 76811-626 76811-628 77298-956
5-Bromo-6-methoxy-1H-pyrrolo[2,3-b]pyridine 97%
5-Bromo-6-methoxy-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₈H₇BrN₂O
MW: 227.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12963483
CAS Number: 1190321-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR