5-Bromo-6-methoxy-1H-indole 95%

Supplier: Ambeed
Warning

A588075-100MG A588075-1G
103828-906EA 300 USD
103828-906 77103-872 77307-896
5-Bromo-6-methoxy-1H-indole 95%
5-Bromo-6-methoxy-1H-indole

Precaución: for Research Use Only

Formula: C₉H₈BrNO
MW: 226.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11848564
CAS Number: 177360-11-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR