5-Bromo-6-methoxy-1H-indazole 97%
Supplier: Ambeed
|
Warning
|
A364215-100MG
A364215-250MG
A364215-1G
A364215-5G
103826-358EA
19
USD
103826-358
103837-376
76788-592
76788-594
76840-606
77306-752
5-Bromo-6-methoxy-1H-indazole 97%
5-Bromo-6-methoxy-1H-indazole
Precaución: for Research Use Only
Formula:
C₈H₇BrN₂O Boiling Pt: 363.4 °C (760 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD11007880 CAS Number: 152626-78-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...