5-Bromo-6-hydroxypicolinic acid 97%

Supplier: Ambeed
Warning

A455353-1G A455353-10G A455353-250MG A455353-5G
103827-840EA 54 USD
103827-840 103838-910 76842-758 77034-142 77034-144 77307-386
5-Bromo-6-hydroxypicolinic acid 97%
5-Bromo-6-hydroxypicolinic acid

Precaución: for Research Use Only

Formula: C₆H₄BrNO₃
MW: 218.0049 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13185807
CAS Number: 1214385-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR