5-Bromo-6-fluoro-1H-indole 96%

Supplier: Ambeed
Warning

A370795-100MG A370795-250MG A370795-5G A370795-1G A370795-10G
103826-466EA 29 USD
103826-466 76989-876 76989-878 76989-880 77306-818 77572-108
5-Bromo-6-fluoro-1H-indole 96%
5-Bromo-6-fluoro-1H-indole

Precaución: for Research Use Only

Formula: C₈H₅BrFN
MW: 214.034 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11848565
CAS Number: 434960-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR