5-Bromo-6-fluoro-1H-indazole 97%
Supplier: Ambeed
|
Warning
|
A300825-250MG
A300825-1G
A300825-5G
A300825-25G
103814-282EA
38
USD
103814-282
103835-432
76772-628
76838-118
76838-120
77305-814
5-Bromo-6-fluoro-1H-indazole 97%
5-Bromo-6-fluoro-1H-indazole
Precaución: for Research Use Only
Formula:
C₇H₄BrFN₂ MW: 215.0225 g/mol Boiling Pt: 332 °C (760 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD11044589 CAS Number: 105391-70-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...