5-Bromo-6-fluoro-1H-benzimidazole 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-6-fluoro-1H-benzo[d]imidazole

A143833-250MG A143833-1G A143833-5G
103803-324EA 68 USD
103803-324 76972-728 77316-628 77316-630
5-Bromo-6-fluoro-1H-benzimidazole 97%
5-Bromo-6-fluoro-1H-benzimidazole

Precaución: for Research Use Only

Formula: C₇H₄BrFN₂
MW: 215.022 g/mol
Boiling Pt: 413.8 °C (760 mm Hg)
Flash Pt: 204.1 °C
Storage Temperature: Ambient
MDL Number: MFCD11846341
CAS Number: 1008360-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR