5-Bromo-6-chloropyridin-3-ol 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-2-chloro-5-hydroxypyridine

A119467-100MG A119467-250MG A119467-25G A119467-1G
103799-768EA 19 USD
103799-768 77136-440 77136-442 77136-444 77297-684
5-Bromo-6-chloropyridin-3-ol 98%
5-Bromo-6-chloropyridin-3-ol

Precaución: for Research Use Only

Formula: C₅H₃BrClNO
MW: 208.45 g/mol
Boiling Pt: 388.6 °C (760 mm Hg)
Melting Pt: 173…177 °C
Flash Pt: 188.824 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09835248
CAS Number: 130115-85-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR