5-Bromo-6-chloronicotinonitrile 98%
Supplier: Ambeed
|
Warning
|
A135867-250MG
A135867-1G
A135867-25G
103802-232EA
18
USD
103802-232
76996-636
77315-104
77315-106
5-Bromo-6-chloronicotinonitrile 98%
5-Bromo-6-chloronicotinonitrile
Precaución: for Research Use Only
Formula:
C₆H₂BrClN₂ Melting Pt: 156…158 °C Flash Pt: 114.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD11840998 CAS Number: 71702-01-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...