5-Bromo-6-chlorobenzo[d][1,3]dioxole 95%

Supplier: Ambeed
Warning

A157982-1G A157982-25G A157982-5G
103805-076EA 25 USD
103805-076 103843-282 77319-126 77319-128 77319-130
5-Bromo-6-chlorobenzo[d][1,3]dioxole 95%
5-Bromo-6-chlorobenzo[d][1,3]dioxole

Precaución: for Research Use Only

Formula: C₇H₄BrClO₂
MW: 235.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12547818
CAS Number: 233770-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR