5-Bromo-6-chloro-1H-indazole 98%

Supplier: Ambeed
Warning

A107842-100MG A107842-250MG A107842-1G A107842-5G
103797-250EA 27 USD
103797-250 103823-018 77008-718 77008-720 77283-646 77283-648
5-Bromo-6-chloro-1H-indazole 98%
5-Bromo-6-chloro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄BrClN₂
Storage Temperature: Ambient
MDL Number: MFCD18381158
CAS Number: 1260382-77-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR