5-Bromo-4-pyrimidinecarboxylic acid 97%

Supplier: Ambeed
Warning

A134225-100MG A134225-10G A134225-250MG A134225-1G A134225-25G A134225-5G
103801-986EA 58 USD
103801-986 77109-426 77109-428 77109-430 77314-710 77314-712 77314-714
5-Bromo-4-pyrimidinecarboxylic acid 97%
5-Bromo-4-pyrimidinecarboxylic acid

Precaución: for Research Use Only

Formula: C₅H₃BrN₂O₂
MW: 202.9900 g/mol
Boiling Pt: 357.2 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09754104
CAS Number: 64224-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR