5-Bromo-4-methylpyrimidine 98%

Supplier: Ambeed
Warning

A189946-10G A189946-250MG A189946-1G A189946-25G A189946-5G
103816-240EA 23 USD
103816-240 103846-836 76721-110 76721-112 77084-616 77084-618 77084-620
5-Bromo-4-methylpyrimidine 98%
5-Bromo-4-methylpyrimidine

Precaución: for Research Use Only

Formula: C₅H₅BrN₂
MW: 173.0100 g/mol
Boiling Pt: 221.3 °C (760 mm Hg)
Flash Pt: 87.663 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10000597
CAS Number: 1439-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR