5-Bromo-4-methylpyridine-2,3-diamine 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2,3-diamino-4-methylpyridine

A104899-250MG A104899-1G A104899-25G
103796-796EA 61 USD
103796-796 103822-522 76780-768 77283-304 77871-404
5-Bromo-4-methylpyridine-2,3-diamine 97%
5-Bromo-4-methylpyridine-2,3-diamine

Precaución: for Research Use Only

Formula: C₆H₈BrN₃
MW: 202.0518 g/mol
Boiling Pt: 331.2 °C (760 mm Hg)
Melting Pt: 161…162 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08062974
CAS Number: 41230-93-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR