5-Bromo-4-methyl-2-nitroaniline 98%

Supplier: Ambeed
Warning

A124123-250MG A124123-25G A124123-1G A124123-5G
103800-484EA 10 USD
103800-484 76870-398 76870-400 77312-616 77312-618
5-Bromo-4-methyl-2-nitroaniline 98%
5-Bromo-4-methyl-2-nitroaniline

Precaución: for Research Use Only

Formula: C₇H₇BrN₂O₂
MW: 231.0500 g/mol
Boiling Pt: 347.9 °C (760 mm Hg)
Melting Pt: 165 °C
Flash Pt: 164.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00156314
CAS Number: 40371-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR