5-Bromo-4-methyl-2-(methylthio)pyrimidine 95%

Supplier: Ambeed
Warning

A111073-100MG A111073-250MG
103797-788EA 75 USD
103797-788 103823-598 77233-570 77233-572
5-Bromo-4-methyl-2-(methylthio)pyrimidine 95%
5-Bromo-4-methyl-2-(methylthio)pyrimidine

Precaución: for Research Use Only

Formula: C₆H₇BrN₂S
MW: 219.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29760803
CAS Number: 1294446-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR