5-bromo-4-methoxypyrimidine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-4-methoxypyrimidine

A185524-100MG A185524-10G A185524-250MG A185524-1G
103815-862EA 39 USD
103815-862 103846-430 76985-882 76985-884 76985-886 77084-068
5-bromo-4-methoxypyrimidine 98%
5-bromo-4-methoxypyrimidine

Precaución: for Research Use Only

Formula: C₅H₅BrN₂O
MW: 189.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00234066
CAS Number: 4319-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR