2-Amino-5-bromo-4-methoxypyridine 95%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-4-methoxypyridin-2-amine

A146895-100MG A146895-250MG
103803-718EA 21 USD
103803-718 103841-812 76984-486 76984-488
2-Amino-5-bromo-4-methoxypyridine 95%
2-Amino-5-bromo-4-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₇BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD15143383
CAS Number: 1232431-11-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR