5-Bromo-4-fluoro-2-methylaniline 98%

Supplier: Ambeed
Warning

A342174-100G A342174-10G A342174-250MG A342174-1G A342174-5G A342174-25G
103825-860EA 9 USD
103825-860 103836-834 76773-156 76773-158 76773-160 76839-870 76839-872 77306-548
5-Bromo-4-fluoro-2-methylaniline 98%
5-Bromo-4-fluoro-2-methylaniline

Precaución: for Research Use Only

Formula: C₇H₇BrFN
Melting Pt: 86…90 °C
Flash Pt: 114.9 °C
Storage Temperature: Ambient
MDL Number: MFCD05865218
CAS Number: 627871-16-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR