5-Bromo-4-fluoro-1H-indole 95%

Supplier: Ambeed
Warning

A579531-100MG A579531-250MG A579531-1G A579531-5G
103828-854EA 40 USD
103828-854 103840-002 76830-764 76830-766 76844-134 76844-136
5-Bromo-4-fluoro-1H-indole 95%
5-Bromo-4-fluoro-1H-indole

Precaución: for Research Use Only

Formula: C₈H₅BrFN
Boiling Pt: 315.9±22.0 °C (760 mm Hg)
Flash Pt: 144.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16609918
CAS Number: 344790-96-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR