5-Bromo-4-fluoro-1H-indazole 95%

Supplier: Ambeed
Warning

A271070-100MG A271070-250MG A271070-1G A271070-5G
103813-034EA 16 USD
103813-034 103852-942 76827-336 76827-338 77092-712 77092-714
5-Bromo-4-fluoro-1H-indazole 95%
5-Bromo-4-fluoro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄BrFN₂
MW: 215.02 g/mol
Boiling Pt: 332.2±22.0 °C (760 mm Hg)
Flash Pt: 154.7 °C
Storage Temperature: Ambient
MDL Number: MFCD11007927
CAS Number: 1082041-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR