5-Bromo-4-cyclopropylpyrimidine 98%

Supplier: Ambeed
Warning

A283290-1G A283290-100MG
103813-608EA 206 USD
103813-608 76837-262 76988-514
5-Bromo-4-cyclopropylpyrimidine 98%
5-Bromo-4-cyclopropylpyrimidine

Precaución: for Research Use Only

Formula: C₇H₇BrN₂
MW: 199.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19688785
CAS Number: 1346697-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR