5-Bromo-4-chloropyrimidine 95%

Supplier: Ambeed
Warning

A180868-1G A180868-25G A180868-5G A180868-100MG A180868-10G A180868-250MG
103807-570EA 16 USD
103807-570 103845-932 77083-442 77083-444 77083-446 77287-196 77287-198 77287-200
5-Bromo-4-chloropyrimidine 95%
5-Bromo-4-chloropyrimidine

Precaución: for Research Use Only

Formula: C₄H₂BrClN₂
Flash Pt: 105.397 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09750158
CAS Number: 56181-39-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR