5-Bromo-4-chloropyridin-3-amine 98%

Supplier: Ambeed
Warning

A149058-10G A149058-250MG A149058-1G A149058-25G A149058-5G
103803-998EA 21 USD
103803-998 103842-118 76871-520 76871-522 77317-594 77317-596 77317-598
5-Bromo-4-chloropyridin-3-amine 98%
5-Bromo-4-chloropyridin-3-amine

Precaución: for Research Use Only

Formula: C₅H₄BrClN₂
Melting Pt: 103…107 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12828485
CAS Number: 89283-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR