5-Bromo-4-chloro-6-methylpyrimidine 95%

Supplier: Ambeed
Warning

A148273-1G A148273-5G
103803-904EA 18 USD
103803-904 103842-020 77317-460 77317-462
5-Bromo-4-chloro-6-methylpyrimidine 95%
5-Bromo-4-chloro-6-methylpyrimidine

Precaución: for Research Use Only

Formula: C₅H₄BrClN₂
MW: 207.46 g/mol
Boiling Pt: 271.2 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00234032
CAS Number: 3438-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR