5-Bromo-4-chloro-2-nitrophenol 95%

Supplier: Ambeed
Danger

A112480-250MG A112480-1G A112480-5G
103798-018EA 29 USD
103798-018 103823-854 77094-050 77310-660 77310-662
5-Bromo-4-chloro-2-nitrophenol 95%
5-Bromo-4-chloro-2-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₃BrClNO₃
MW: 252.45 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22383651
CAS Number: 855400-82-7
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR