5-Bromo-4-chloro-2-methylpyrimidine 97%

Supplier: Ambeed
Warning

A143421-100MG A143421-250MG A143421-1G A143421-10G
103803-280EA 72 USD
103803-280 76972-700 76972-702 77316-546 77671-044
5-Bromo-4-chloro-2-methylpyrimidine 97%
5-Bromo-4-chloro-2-methylpyrimidine

Precaución: for Research Use Only

Formula: C₅H₄BrClN₂
MW: 207.45 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11046966
CAS Number: 861383-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR